
3,5-Difluorobenzylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:90390-27-5
:
Formula:C₇H₇F₂N
MDL Number:MFCD00061244
:143.14 g/mol
Boiling Pt:184 °C
Density:1.21 g/cm³ (25 °C)
Flash Pt:73 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together