
2,4-Difluorobenzoic acid 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1583-58-0
:
Formula:C₇H₄F₂O₂
MDL Number:MFCD00011670
:158.1 g/mol
Melting Pt:184…190 °C
EINECS:216-430-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together