


2-(Trifluoromethyl)-p-anisidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:53903-49-4
:
Formula:C₈H₈F₃NO
MDL Number:MFCD04972660
:191.15 g/mol
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Frequently Bought Together