
2,6-Dichloro-4-(trifluoromethyl)nicotinamide 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:158063-67-3
:
Formula:C₇H₃Cl₂F₃N₂O
MDL Number:MFCD00173903
:259.02 g/mol
Melting Pt:194…200 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together