Order Entry
Czech Republic
ContactUsLinkComponent
2,3-Dichloro-5-(trifluoromethyl)pyridine 98%
Corrosive
Irritant
Environment
2,3-Dichloro-5-(trifluoromethyl)pyridine 98%
Catalog # APOSPC2556D-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
Environment
2,3-Dichloro-5-(trifluoromethyl)pyridine 98%
Catalog # APOSPC2556D-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :215.99 g/mol
Boiling Pt:79…81 °C/20mmHg
Density:1.549 g/cm³ (25 °C)
Flash Pt:79 °C
Storage Temperature:Ambient
EINECS:410-340-5
UN:3082
ADR:9+EHS,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:69045-84-7
Formula:C₆H₂Cl₂F₃N
MDL Number:MFCD00042243