
2-Bromo-5-(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1483-55-2
:
Formula:C₈H₃BrF₃N
MDL Number:MFCD03412187
Boiling Pt:107…115 °C/10mmHg
Melting Pt:50…51 °C
UN:3439
ADR:6.1,III
:250.02 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together