
2,4-Dichloro-5-fluorobenzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:86522-89-6
:
Formula:C₇H₃Cl₂FO₂
MDL Number:MFCD00075281
:209 g/mol
Melting Pt:144…146 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together