
2,6-Difluoro-4-methoxybenzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:123843-65-2
:
Formula:C₈H₆F₂O₃
MDL Number:MFCD02949623
:188.13 g/mol
Melting Pt:184…188 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together