1,2-Dibromopentafluoropropyl-2,2,2,3,3-pentafluoropropyl ether 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:396716-50-0
:
Formula:C₆H₂Br₂F₁₀O
MDL Number:MFCD03094147
Storage Temperature:Ambient
Frequently Bought Together