Order Entry
Czech Republic
ContactUsLinkComponent
 
Barium hexafluorosilicate 98%
Catalog # APOSPC2147-50G
Supplier:  Apollo Scientific
CAS Number:  
Barium hexafluorosilicate 98%
Catalog # APOSPC2147-50G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:17125-80-3
Formula:BaF₆Si
MDL Number:MFCD00014185