Order Entry
Czech Republic
ContactUsLinkComponent
3-Chloro-5-(trifluoromethyl)pyridine 97%
Irritant
3-Chloro-5-(trifluoromethyl)pyridine 97%
Catalog # APOSPC2075G-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Chloro-5-(trifluoromethyl)pyridine 97%
Catalog # APOSPC2075G-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :181.55 g/mol
Boiling Pt:144 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:85148-26-1
Formula:C6H3ClF3N
MDL Number:MFCD00042227