
2-Iodo-5-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:105202-02-6
:
Formula:C₇H₅F₃IN
MDL Number:MFCD04972658
:287.0210 g/mol
Boiling Pt:102…104 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together