Order Entry
Czech Republic
ContactUsLinkComponent
 
2-Bromo-3-fluoro-4-methylpyridine 97%
Catalog # APOSPC201348-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-3-fluoro-4-methylpyridine 97%
Catalog # APOSPC201348-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :190.01 g/mol
Storage Temperature:Ambient
CAS Number:884494-37-5
Formula:C₆H₅BrFN
MDL Number:MFCD04112499