
2-[4-(Trifluoromethyl)phenyl]ethylamine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:775-00-8
:
Formula:C₉H₁₀F₃N
MDL Number:MFCD01529875
:189.181 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together