3-Methoxy-4-(trifluoromethyl)phenylamine 98%
Catalog # APOSPC201016-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:191.15 g/mol
Storage Temperature:Ambient
CAS Number:106877-20-7
Formula:C₈H₈F₃NO
MDL Number:MFCD11840978