
Ethyl-4-chloro-5,7-difluoroquinoline-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:311346-69-7
:
Formula:C₁₂H₈ClF₂NO₂
MDL Number:MFCD00173416
:271.65 g/mol
Melting Pt:55…57 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together