3-(3-Fluorophenoxy)propionic acid 95+%
Catalog # APOSPC200561-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.1644 g/mol
Melting Pt:92…96 °C
Storage Temperature:Ambient
CAS Number:133077-42-6
Formula:C₉H₉FO₃
MDL Number:MFCD01931913