Ethyl-3-(4-fluorophenyl)-5-methylisoxazole-4-carboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:954230-39-8
:
Formula:C₁₃H₁₂FNO₃
MDL Number:MFCD09756447
Melting Pt:58…59 °C
:249.24 g/mol
Storage Temperature:Ambient
Frequently Bought Together