Order Entry
Czech Republic
ContactUsLinkComponent
Diethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]-1H-1,2,3-triazole-4,5-dicarboxylate
Diethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]-1H-1,2,3-triazole-4,5-dicarboxylate
Catalog # APOSPC200482-1G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]-1H-1,2,3-triazole-4,5-dicarboxylate
Catalog # APOSPC200482-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :349.32 g/mol
Storage Temperature:Ambient
CAS Number:1610376-99-2
Formula:C₁₆H₁₆FN₃O₅
MDL Number:MFCD28023527