Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
3-(4-Chloro-2-fluorophenyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one
Catalog # APOSPC200359-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(4-Chloro-2-fluorophenyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one
Catalog # APOSPC200359-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :284.72 g/mol
Melting Pt:96…97 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1335220-63-7
Formula:C₁₃H₁₄ClFN₂O₂
MDL Number:MFCD20232921