4-Methyl-2-[4-(trifluoromethyl)phenyl]benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:273727-19-8
:
Formula:C₁₅H₁₁F₃O₂
MDL Number:MFCD18425410
:280.25 g/mol
Storage Temperature:Ambient
Frequently Bought Together