Order Entry
Czech Republic
ContactUsLinkComponent
5-Chloro-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile
5-Chloro-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile
Catalog # APOSPC200326-500MG
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile
Catalog # APOSPC200326-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :262.62 g/mol
Melting Pt:240 °C
Storage Temperature:Ambient
CAS Number:1092352-56-1
Formula:C₁₀H₆ClF₃N₂O
MDL Number:MFCD11553041