Methyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-7-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1186405-14-0
:
Formula:C₁₀H₆ClF₃N₂O₂
MDL Number:MFCD12827781
Melting Pt:97…98 °C
Storage Temperature:Ambient
Frequently Bought Together