Methyl-2-amino-3-chloro-5-(trifluoromethyl)isonicotinate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1171919-10-0
:
Formula:C₈H₆ClF₃N₂O₂
MDL Number:MFCD12498715
Melting Pt:131…132 °C
:254.6 g/mol
Storage Temperature:Ambient
Frequently Bought Together