2,3-Dichloro-5-(trifluoromethyl)isonicotinic acid
Catalog # APOSPC200277-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:260 g/mol
Melting Pt:192…193 °C
Storage Temperature:Ambient
CAS Number:1135283-33-8
Formula:C₇H₂Cl₂F₃NO₂
MDL Number:MFCD12025872