

4-Fluoro-2-methoxypyridine
Catalog # APOSPC200266-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:127.12 g/mol
Boiling Pt:22…23 °C/2mmHg
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:96530-81-3
Formula:C₆H₆FNO
MDL Number:MFCD09835267