
2,4-Dichloro-5-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:888327-38-6
:
Formula:C₆H₂Cl₂F₃N
MDL Number:MFCD14525480
:215.988 g/mol
Boiling Pt:76 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together