1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]azetidin-3-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1311798-09-0
:
Formula:C₉H₈ClF₃N₂O
MDL Number:MFCD18266134
:252.63 g/mol
Melting Pt:88…90 °C
Storage Temperature:Ambient
Frequently Bought Together