2,3-Dichloro-5-(trifluoromethyl)isonicotinoyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1198475-32-9
:
Formula:C₇HCl₃F₃NO
MDL Number:MFCD14581644
:278.45 g/mol
Boiling Pt:75 °C/0.01mmHg
Storage Temperature:Ambient
Frequently Bought Together