3-Nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1186405-07-1
:
Formula:C₉H₄F₃N₃O₄
MDL Number:MFCD12827808
:275.15 g/mol
Melting Pt:172 °C (dec.)
Storage Temperature:Ambient
Frequently Bought Together