7-Hydroxy-2-methyl-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1198475-25-0
:
Formula:C₉H₅F₃N₄O
MDL Number:MFCD14581631
:242.16 g/mol
Melting Pt:302…304 °C
Storage Temperature:Ambient
Frequently Bought Together