4,6-Bis(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:89990-37-4
:
Formula:C₈H₃F₆N₃O
MDL Number:MFCD12827802
:271.12 g/mol
Melting Pt:>290 °C
Storage Temperature:Ambient
Frequently Bought Together