
1,3-Difluoro-2-iodobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13697-89-7
:
Formula:C₆H₃F₂I
MDL Number:MFCD04039247
:239.99 g/mol
Boiling Pt:71 °C/14mmHg
Melting Pt:24 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together