Order Entry
Czech Republic
ContactUsLinkComponent
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine 95%
Irritant
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine 95%
Catalog # APOSPC1931E-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine 95%
Catalog # APOSPC1931E-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Melting Pt:159…161 °C
 :197.541 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:76041-71-9
Formula:C₆H₃ClF₃NO
MDL Number:MFCD00153095