
6-Chloro-7-hydroxy-4-(trifluoromethyl)coumarin 97%
Catalog # APOSPC1931C-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
Melting Pt:178…182 °C
:264.59 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:119179-66-7
Formula:C₁₀H₄ClF₃O₃
MDL Number:MFCD00068095