Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluorobenzylmagnesium chloride
2-Fluorobenzylmagnesium chloride 0,25 M in diethyl ether
2-Fluorobenzylmagnesium chloride 0,25 M in diethyl ether
APOSPC1897-100ML
:
Apollo Scientific
:
2-Fluorobenzylmagnesium chloride 0,25 M in diethyl ether
APOSPC1897-100ML
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mL
Seq:
2440
:
:
168.878 g/mol
:
Ambient
:
1993
:
3,III
:
GHS02
:
GHS05
:
GHS07
:
120608-58-4
:
C₇H₆ClFMg
:
MFCD01319900