
5-(Trifluoromethyl)-m-anisidine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:349-55-3
:
Formula:C₈H₈F₃NO
MDL Number:MFCD00000393
:191.15 g/mol
Melting Pt:50…52 °C
Flash Pt:113 °C
EINECS:206-487-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together