
6-(Trifluoromethyl)nicotinamide 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:386715-35-1
:
Formula:C₇H₅F₃N₂O
MDL Number:MFCD01862646
:190.1200 g/mol
Melting Pt:185…187 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together