

2-Chloro-1,1-difluoroethylene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:359-10-4
:
Formula:C₂HClF₂
:98.48
Boiling Pt:–18 °C
Melting Pt:–138 °C
UN:1954
ADR:2.1,
:GHS04
:GHS07
Storage Temperature:Ambient
Frequently Bought Together