Order Entry
Czech Republic
ContactUsLinkComponent
4-Methoxy-3-(trifluoromethyl)phenylboronic acid 98%
Irritant
4-Methoxy-3-(trifluoromethyl)phenylboronic acid 98%
Catalog # APOSPC1772-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Methoxy-3-(trifluoromethyl)phenylboronic acid 98%
Catalog # APOSPC1772-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.95 g/mol
Melting Pt:198…202 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:149507-36-8
Formula:C₈H₈BF₃O₃
MDL Number:MFCD07363790