
6-(Trifluoromethyl)oxindole 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1735-89-3
:
Formula:C₉H₆F₃NO
MDL Number:MFCD01075220
:201.15 g/mol
Melting Pt:184…188 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together