
2-Fluoro-5-(trifluoromethyl)phenylboronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:352535-96-7
:
Formula:C₇H₅BF₄O₂
MDL Number:MFCD03701538
:207.92 g/mol
Melting Pt:104…109 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together