
2-Bromo-5-(trifluoromethoxy)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:887267-47-2
:
Formula:C₇H₅BrF₃NO
MDL Number:MFCD08458013
:256.02 g/mol
Density:1.71 g/cm³ (25 °C)
Flash Pt:97 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together