Order Entry
Czech Republic
ContactUsLinkComponent
1-Bromo-2,3,5,6-tetrafluorobenzene 98%
Flammable
Irritant
1-Bromo-2,3,5,6-tetrafluorobenzene 98%
Catalog # APOSPC1560-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
1-Bromo-2,3,5,6-tetrafluorobenzene 98%
Catalog # APOSPC1560-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :228.97 g/mol
Boiling Pt:142…144 °C
Density:1.883 g/cm³ (25 °C)
Flash Pt:53 °C
Storage Temperature:Ambient
EINECS:216-327-9
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:1559-88-2
Formula:C₆HBrF₄
MDL Number:MFCD00000308