
4-[5-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:871252-64-1
:
Formula:C₁₃H₈F₃NO
MDL Number:MFCD06797372
:251.21 g/mol
Melting Pt:92…94 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together