Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
2-Chloro-3-fluoro-5-methylpyridine
Catalog # APOSPC1401-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Chloro-3-fluoro-5-methylpyridine
Catalog # APOSPC1401-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :145.56 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:34552-15-3
Formula:C₆H₅ClFN
MDL Number:MFCD06658238