
1-(2-Bromo-4,6-difluorophenoxy)-2-chloroethane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175203-19-7
:
Formula:C₈H₆BrClF₂O
MDL Number:MFCD00068037
:271.5 g/mol
Boiling Pt:119…122 °C/15mmHg
:GHS07
Storage Temperature:Ambient
Frequently Bought Together