
Diethyl (bromodifluoromethyl)phosphonate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:65094-22-6
:
Formula:C₅H₁₀BrF₂O₃P
MDL Number:MFCD00069118
:267.01 g/mol
Boiling Pt:40…41 °C/0.5mmHg
Density:1.503 g/cm³ (25 °C)
Flash Pt:>113 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together