
trans-1,2-Bis(perfluorohexyl)ethylene 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51249-67-3
:
Formula:C₁₄H₂F₂₆
MDL Number:MFCD03094106
Boiling Pt:195 °C
Density:1.766 g/cm³ (25 °C)
:664.13 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together