Order Entry
Czech Republic
ContactUsLinkComponent
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene 97%
Irritant
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene 97%
Catalog # APOSPC1228B-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene 97%
Catalog # APOSPC1228B-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :410.16 g/mol
Boiling Pt:209 °C
Density:1.659 g/cm³ (25 °C)
Flash Pt:97 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:802-93-7
Formula:C₁₂H₆F₁₂O₂
MDL Number:MFCD00042090